Diethylzinc 15% (w/w) 1 M in hexane, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
20551-1001
205518000
TS20551-1001EA
135
USD
TS20551-1001
TS205518000
Diethylzinc 15% (w/w) 1 M in hexane, AcroSeal™
Diethylzinc
Formula:
C₄H₁₀Zn Melting Pt: 39…28 °C Flash Pt: –40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009021 CAS Number: 557-20-0 EINECS: 209-161-3 UN: 3394 ADR: 4.2, |
Specification Test Results
Appearance | Clear to hazy colorless to yellow to gray-brown Solution |
Titration Complexometric | 0.85 to 1.00 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...