4-Carboxyphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid
34440-0250
34440-0010
34440-0050
TS34440-0250EA
529
USD
TS34440-0250
TS34440-0010
TS34440-0050
4-Carboxyphenylboronic acid 97%
4-Carboxyphenylboronic acid
Formula:
C₇H₇BO₄ Melting Pt: 238…240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151801 CAS Number: 14047-29-1 |
Specification Test Results
Appearance | White to light yellow to light beige Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...