n-Dodecyl-β-D-maltoside 99%, high_purity

Supplier: Thermo Scientific Chemicals

Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside

32937-0050 32937-0010 32937-0250
TS32937-0050EA 672 USD
TS32937-0050 TS32937-0010 TS32937-0250
n-Dodecyl-β-D-maltoside 99%, high_purity
n-Dodecyl-β-D-maltoside
Formula: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
Melting Pt: 217 °C
Storage Temperature: Freezer
MDL Number: MFCD00043012
CAS Number: 69227-93-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥98.5 % (ELSD)
Specific optical rotation +45° to +49.5° (20°C, 589 nm) (c=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR