TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,3',3''-Phosphinetriyltripropanoic acid hydrochloride, TCEP, Tris(2-carboxyethyl)phosphine, hydrochloride
36383-0100
36383-0010
TS36383-0100EA
647
USD
TS36383-0100
TS36383-0010
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98%
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)
Formula:
C₉H₁₆ClO₆P MW: 286.65 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00145469 CAS Number: 51805-45-9 UN: 3261 ADR: 8 II |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Solubility | (50mg/ml water) Clear colorless |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...