Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine

13275-0050 13275-0010
TS13275-0050EA 201 USD
TS13275-0050 TS13275-0010
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) 97%
Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine)
Formula: C₁₀H₂₆N₄
MW: 202.35 g/mol
Boiling Pt: 130 °C (.1 mmHg)
Melting Pt: 26…30 °C
Density: 0.937 g/cm³
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008215
CAS Number: 71-44-3
EINECS: 200-754-2
UN: 3259
ADR: 8 II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or low melting mass
Infrared spectrum Conforms
Melting point 26 to 30 °C
Water ≤3 % (K.F.)
Titration with HClO4 ≥96.0 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR