Sodium-tert-butanolate 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

25255-5000 AC25255-0025 AC25255-0050 AC25255-1000
TS25255-5000EA 217 USD
TS25255-5000 TS25255-0025 TS25255-0050 TS25255-1000
Sodium-tert-butanolate 99%, pure
Sodium tert-butanolate
Formula: C₄H₉NaO
Melting Pt: 180 °C
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00040575
CAS Number: 865-48-5
EINECS: 212-741-9
UN: 3206
ADR: 4.2,II

Order Now

Specification Test Results

Appearance White to light brown Powder
Titration with HCl 98.5 to 101.5 %
NaOH + Na2CO3 (as NaOH) ≤1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR