Anthrone ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Anthracen-9(10H)-one
10496-1000
10496-0250
TS10496-1000EA
264
USD
TS10496-1000
TS10496-0250
Anthrone ACS
Anthrone
Formula:
C₁₄H₁₀O MW: 194.23 g/mol Boiling Pt: 721 °C (760 mmHg) Melting Pt: 152…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001187 CAS Number: 90-44-8 EINECS: 201-994-0 |
Specification Test Results
Appearance | Yellow Crystals |
Infrared spectrum | Conforms |
Melting point | 152 to 157 °C |
Solubility | (in ethyl acetate) Passes test |
Absorbance | ≤0.045 (620 nm, 1 cm cell) |
Sensitivity | (to carbohydrates) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...