Benzamidine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzimidamide hydrochloride, Benzamidinium chloride, Phenylamidine hydrochloride, Benzene-1-carboximidamide hydrochloride

40179-5000 40179-0050 40179-0250 40179-1000
TS40179-5000EA 1312 USD
TS40179-5000 TS40179-0050 TS40179-0250 TS40179-1000
Benzamidine hydrochloride 99%
Benzamidine hydrochloride
Formula: C₇H₉ClN₂
Melting Pt: 167…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00066285
CAS Number: 1670-14-0
EINECS: 216-795-4

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR