Copper (II) ethylenediamine complex 1 M in water

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: CupriethylendiamineBis(ethylenediamine)copper dihydroxideCopper(II)-ethylenediame complex

258550025 25855-0010
TS25855-0010EA 91.5 USD
TS25855-0010 TS258550025
Copper (II) ethylenediamine complex 1 M in water
Copper (II) ethylenediamine
Formula: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MW: 217.76 g/mol
Density: 1.1 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00274628
CAS Number: 14552-35-3
UN: 1761
ADR: 8,II

Order Now

Specification Test Results

Appearance Dark blue Solution
Solubility (1.3 g cellulose + 25ml water + 25ml CED complex): clear
Ratio (Ethylenediamine to copper) 1.90:1 to 2.10:1
Strength 0.98 to 1.02 M (Copper molarity)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR