Acrylonitrile 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14963-0010
14963-0025
14963-0100
14963-1000
TS14963-0010EA
55
USD
TS14963-0010
TS14963-0025
TS14963-0100
TS14963-1000
Acrylonitrile 99+%
Acrylonitrile
Formula:
C₃H₃N Boiling Pt: 77.3 °C (760 mmHg) Melting Pt: –83.5 °C Density: 0.8 g/cm³ Flash Pt: –0.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001927 CAS Number: 107-13-1 EINECS: 203-466-5 UN: 1093 ADR: 3,I Merck Index: 13,00133 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3900 to 1.3920 (20°C, 589 nm) |
Water | ≤0.5 % |
GC | ≥99.0 % |
Color scale | ≤15 APHA |
Stabilizer | 35 to 45 ppm MEHQ |
Non-volatile matter | ≤100 ppm |
pH | 5.9 to 7.6 (5 % in water) |
Copper (Cu) | ≤0.1 ppm |
Iron (Fe) | ≤0.1 ppm |
Impurity | ≤100 ppm Oxazole |
Acidity (CH3COOH) | ≤20 ppm |
Aldehyde (C2H4O) | ≤50 ppm |
Acetone | ≤100 ppm |
Peroxides | ≤0.2 ppm (as H2O2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...