N-Ethylmaleimide (NEM) 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione
15610-0050
15610-0500
15610-2500
15610-0100
TS15610-0050EA
79
USD
TS15610-0050
TS15610-0500
TS15610-2500
TS15610-0100
N-Ethylmaleimide (NEM) 99+%
N-Ethylmaleimide
Formula:
C₆H₇NO₂ MW: 125.13 g/mol Boiling Pt: 210 Melting Pt: 46 Flash Pt: 73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005509 CAS Number: 128-53-0 EINECS: 204-892-4 UN: 2928 ADR: 6.1 II Merck Index: 13,03855 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 41 to 45 °C |
Solubility | (5% in 100% EtOH) Clear colorless to almost colorless solution |
HPLC | ≥99.0 % |
Residue after ignition | ≤0.01 % |
Aluminium (Al) | ≤1 ppm |
Antimony (Sb) | ≤1 ppm |
Arsenic (As) | ≤1 ppm |
Boron (B) | ≤1 ppm |
Calcium (Ca) | ≤5 ppm |
Chromium (Cr) | ≤1 ppm |
Copper (Cu) | ≤1 ppm |
Iron (Fe) | ≤1 ppm |
Lead (Pb) | ≤1 ppm |
Magnesium (Mg) | ≤1 ppm |
Manganese (Mn) | ≤1 ppm |
Nickel (Ni) | ≤1 ppm |
Potassium (K) | ≤20 ppm |
Sodium (Na) | ≤20 ppm |
Zinc (Zn) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...