Diethylenetriaminepentaacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DTPAIdranal VDiethylenetriamine-N,N,N',N'',N''-pentaacetic acid2,2',2'',2'''-((((Carboxymethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetic acidDiethylenetriaminepentaacetic acidN-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid)Kestranal® Aza PaTitriplex® V

11432-0010 11432-0050 11432-2500
TS11432-0050EA 389 USD
TS11432-0050 TS11432-2500 TS11432-0010
Diethylenetriaminepentaacetic acid 98+%
Diethylenetriaminepentaacetic acid
Formula: C₁₄H₂₃N₃O₁₀
MW: 393.35 g/mol
Melting Pt: 220 °C (dec.)
Density: 1.56 g/cm³
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00004289
CAS Number: 67-43-6
EINECS: 200-652-8
Merck Index: 12,07266

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration excess NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR