Trifluoroacetic anhydride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trifluoracetic anhydrideTFAA

147810250 147812501 14781-5000 147810025 14781-1000
TS147812501EA 155 USD
TS147812501 TS147810250 TS14781-5000 TS147810025 TS14781-1000
Trifluoroacetic anhydride 99+%
Trifluoroacetic anhydride
Formula: C₄F₆O₃
MW: 210.03 g/mol
Boiling Pt: 39.1 °C (760 mmHg)
Melting Pt: –63.5 °C
Density: 1.51 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000416
CAS Number: 407-25-0
EINECS: 206-982-9
UN: 3265
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥99.0 %
Chloride (Cl) ≤50 ppm
Fluoride (F) ≤50 ppm
Sulfate (SO4) ≤500 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR