1,10-Phenanthroline 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Phenanthroline

15753-0250 15753-0050 15753-1000 15753-5000
TS15753-0250EA 128 USD
TS15753-0250 TS15753-0050 TS15753-1000 TS15753-5000
1,10-Phenanthroline 99+%
1,10-Phenanthroline
Formula: C₁₂H₈N₂
MW: 180.21 g/mol
Boiling Pt: 360
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00011678
CAS Number: 66-71-7
EINECS: 200-629-2
UN: 2811
ADR: 6.1 III
Merck Index: 12,07356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5092a8ec81301ed6ed30f75429f9f27784db47a5&vid=106c4325bb7c310de902d3118cf4b31bcee3dc92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light yellow to light pink Powder
Infrared spectrum Conforms
Melting point 114 to 120 °C
Sulfated ash ≤0.1 %
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR