N,N-Dimethylformamide-[D₇] 99.5 atom % D for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N,N-Di[2H3]methyl[2H]formamide
, DMF-D7
, DMF-d7
, N,N-Dimethylformamide-[²H₇]
, Formic acid dimethylamide-D7
, Heptadeutero-N,N-dimethylformamide
, Formdimethylamide-D7
, N,N-Dimethylformamide-d7
32073-0075
TS32073-0075EA
407
USD
TS32073-0075
N,N-Dimethylformamide-[D₇] 99.5 atom % D for NMR spectroscopy
N,N-Dimethylformamide-[D7]
Formula:
DCOON(CD₃)₂ MW: 80.04 g/mol Boiling Pt: 152 °C (1013 hPa) Melting Pt: –60 °C Density: 1.03 g/cm³ Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003286 CAS Number: 4472-41-7 EINECS: 224-745-8 UN: 2265 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.1 % (Coulometric) |
D-Enrichment | ≥99.4 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...