N,N-Dimethylformamide-[D₇] 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

18360-0100 18360-0010 183600050 183600250
TS18360-0100EA 522 USD
TS18360-0100 TS18360-0010 TS183600050 TS183600250
N,N-Dimethylformamide-[D₇] 99.5 atom % D for NMR spectroscopy
N,N-Dimethylformamide-[D7]
Formula: DCOON(CD₃)₂
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1.03 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003286
CAS Number: 4472-41-7
EINECS: 224-745-8
UN: 2265
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
GC ≥99.0 %
D-Enrichment ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR