Molecular sieve 13X (1.0 nm, 10 Å) 4 - 8 mesh
Supplier: Thermo Scientific Chemicals
19730-0050
197305000
TS19730-0050EA
358
USD
TS19730-0050
TS197305000
Molecular sieve 13X (1.0 nm, 10 Å) 4 - 8 mesh
Molecular sieve 13X (1.0 nm, 10 Å)
MDL Number:
MFCD00164506 CAS Number: 63231-69-6 |
Specification Test Results
Appearance | Beige to light brown Beads |
Particle size | 4 to 8 mesh |
Bulk density | ≥600 g/l |
Residual water | ≤1.5 % (550°C, 2 h) |
CO2 adsorption capacity | ≥11.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...