Tetrabutylammonium hydroxide 40% (w/w) 1.5 M in water

Supplier: Thermo Scientific Chemicals
Danger

17661-2500 176610010 176610025 176610500
TS17661-2500EA 301 USD
TS17661-2500 TS176610010 TS176610025 TS176610500
Tetrabutylammonium hydroxide 40% (w/w) 1.5 M in water
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
Density: 0.995 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤150 APHA
Titration with HCl 38 to 42 %
Solubility (5% in water) Clear colorless
Specific optical rotation +20.0° to +24.0° (20°C, 589 nm) (c=1%, water)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR