Perchloric acid 0.1 M in acetic acid, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
34863-0010
348630025
TS34863-0010EA
102
USD
TS34863-0010
TS348630025
Perchloric acid 0.1 M in acetic acid, pure
Perchloric acid
Formula:
HClO₄ Density: 1.06 g/cm³ Flash Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 EINECS: 231-512-4 UN: 2920 ADR: 8,II |
|
Specification Test Results
Appearance | Clear colorless Solution |
Water | ≤0.2 % (K.F.) |
Strength | 0.0998 to 0.1002 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...