Sodium methoxide 0.5 M in methanol, AcroSeal™ ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium methylate, Sodium methanolate

44588-8000 AC44588-1000
TS44588-8000EA 283 USD
TS44588-8000 TS44588-1000
Sodium methoxide 0.5 M in methanol, AcroSeal™ ACS
Sodium methoxide
Formula: CH₃NaO
MW: 54.02 g/mol
Boiling Pt: 65 °C (760 mmHg)
Melting Pt: 127
Density: 0.8 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
UN: 1992
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl 0.48 to 0.52 M
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR