Sodium methoxide 0.5 M in methanol, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium methylate, Sodium methanolate

42722-1000 42722-8000
TS42722-1000EA 92 USD
TS42722-1000 TS42722-8000
Sodium methoxide 0.5 M in methanol, AcroSeal™
Sodium methoxide
Formula: CH₃NaO
MW: 54.02 g/mol
Melting Pt: 127
Density: 0.8 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
UN: 1992
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.45 to 0.60 M
UV at 240 nm A: ≤0.02
Residue after evaporation ≤2 ppm
Acidity ≤0.00015 meq/g
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR