Magnesium sulfate, anhydrous 97%, pure

Supplier: Thermo Scientific Chemicals

41348-0250 41348-5000 AC41348-0025 AC41348-0050
TS41348-0250EA 1333 USD
TS41348-0250 TS41348-5000 TS41348-0025 TS41348-0050
Magnesium sulfate, anhydrous 97%, pure
Magnesium sulphate
Formula: MgSO₄
Melting Pt: 1124 °C
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light gray Granular powder (May contain gray particles)
Titration Complexometric ≥96.0 %
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤100 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm
Silicon dioxide (SiO2) ≤1000 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR