Magnesium oxide 98%, powder Ø 99% <150 µm (-100 mesh), Extra Pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Magnesia usta
AC26383-0100
26383-5000
AC26383-0025
TS26383-5000EA
98
USD
TS26383-5000
TS26383-0100
TS26383-0025
Magnesium oxide 98%, powder Ø 99% <150 µm (-100 mesh), Extra Pure
Magnesium oxide
Formula:
MgO MW: 40.3 g/mol Boiling Pt: 3600 °C (760 mmHg) Melting Pt: 2800 °C Density: 3.58 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 |
Specification Test Results
Appearance | White Powder |
Titration Complexometric | ≥97.5 % |
Sulfate (SO4) | ≤2500 ppm |
Calcium oxide (CaO) | ≤3000 ppm |
Silicon dioxide (SiO2) | ≤500 ppm |
Loss on ignition | ≤2 % |
Aluminium oxide (Al2O3) | ≤400 ppm |
Iron(III)oxide (Fe2O3) | ≤400 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...