Magnesium oxide 99.99% (trace metals basis), extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesia usta

19343-0500 AC19343-0100
TS19343-0500EA 240 USD
TS19343-0500 TS19343-0100
Magnesium oxide 99.99% (trace metals basis), extra pure
Magnesium oxide
Formula: MgO
Boiling Pt: 3600 °C (760 mmHg)
Melting Pt: 2800 °C
Density: 3.58 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Specification Test Results

Appearance White Powder
Total trace metal impurities ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR