Zinc acetate 99%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
37008-1000
AC37008-0250
TS37008-1000EA
288
USD
TS37008-1000
TS37008-0250
Zinc acetate 99%, pure
Zinc acetate
Formula:
C₄H₆O₄Zn MW: 183.48 Boiling Pt: 908 Melting Pt: 83…86 °C Density: 1.735 Storage Temperature: Ambient |
MDL Number:
MFCD00012454 CAS Number: 557-34-6 EINECS: 209-170-2 UN: 3077 ADR: 9 III Merck Index: 12,10256 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Complexometric | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...