D-(+)-Xylose 99+%
Supplier: Thermo Scientific Chemicals
14100-0025
14100-1000
14100-5000
TS14100-0025EA
594
USD
TS14100-0025
TS14100-1000
TS14100-5000
D-(+)-Xylose 99+%
D(+)-Xylose
Formula:
C₅H₁₀O₅ Melting Pt: 148…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151475 CAS Number: 58-86-6 EINECS: 200-400-7 Merck Index: 13,10142 |
|
Specification Test Results
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.2 % (K.F.) |
Sulfated ash | ≤0.2 % |
HPLC | ≥99.0 % (ELSD) |
Specific optical rotation | +18.0° to +21.0° (20°C, 589 nm) (c=10, H2O/NH3) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...