Calcium oxide 96%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: LimeQuicklimecausticCalcium oxide from marble

AC19691-0010 19691-0025
TS19691-0025EA 254 USD
TS19691-0025 TS19691-0010
Calcium oxide 96%, Extra Pure
Calcium oxide
Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850 °C (1013 hPa)
Melting Pt: 2570 °C
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
UN: 1910
ADR: 8,III
Merck Index: 13,01691

Order Now

Specification Test Results

Appearance White to light beige Powder
Titration Complexometric ≥95 %
Arsenic (As) ≤3 ppm
Lead (Pb) ≤10 ppm
Insoluble matter ≤0.5 % (in HCl)
Loss on ignition ≤3.5 % (800°C)
Mg and alkali salts (SO4) ≤2.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR