Dimidium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide

21534-0010 21534-2500
TS21534-0010EA 302 USD
TS21534-0010 TS21534-2500
Dimidium bromide 98%
Dimidium bromide
Formula: C₂₀H₁₈BrN₃
Melting Pt: 230 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011757
CAS Number: 518-67-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e669239c79726ce91f44ef372cbfed426e90f93b&vid=74b03720866852364cb9050de861a4f7c0e5e57b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Dark red to purple Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 % (on dry substance)
Water ≤5 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR