tert-Butanol 99.5%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

10771-0010 107710025 107710100
TS10771-0010EA 121 USD
TS10771-0010 TS107710025 TS107710100
tert-Butanol 99.5%, extra pure
tert-Butanol
Formula: (CH₃)₃COH
Boiling Pt: 83 °C (760 mmHg)
Melting Pt: 25…25.5 °C
Density: 0.78 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II
Merck Index: 12,01577
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid after melting
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3880 (20°C, 589 nm)
Water ≤0.03 % (K.F.)
GC ≥99.4 %
Residue after evaporation ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR