N-Ethyldiisopropylamine 99.5+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA, Ethyldiisopropylamine

36784-0010 36784-0025 36784-1000 36784-0250 36784-0500
TS36784-0010EA 482 USD
TS36784-0010 TS36784-0025 TS36784-1000 TS36784-0250 TS36784-0500
N-Ethyldiisopropylamine 99.5+%
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
Boiling Pt: 127 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0.756 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
EINECS: 230-392-0
UN: 3384
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4150 (20°C, 589 nm)
Water ≤0.3 %
GC ≥99.50 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR