N-Ethyldiisopropylamine 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA, Ethyldiisopropylamine

11522-1000 11522-5000 11522-0026 11522-0101 11522-0250
TS11522-1000EA 88 USD
TS11522-1000 TS11522-5000 TS11522-0026 TS11522-0101 TS11522-0250
N-Ethyldiisopropylamine 99+%
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 127
Melting Pt: -50
Density: 0.756 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
EINECS: 230-392-0
UN: 3384
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4150 (20°C, 589 nm)
Water ≤0.30 % (K.F.)
GC ≥99.0 %
Diisopropylamine ≤0.30 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR