1-Hexanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium hexane-1-sulfonate, Sodium hexane-1-sulphonate, Sodium hexanesulfonate, Sodium hexanesulphonate, Sodium 1-hexanesulfonate, Sodium 1-hexanesulphonate

39667-1000 AC39667-0250
TS39667-1000EA 1092 USD
TS39667-1000 TS39667-0250
1-Hexanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
1-Hexanesulphonic acid sodium salt
Formula: C₆H₁₃O₃SNa
MW: 188.22 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00007542
CAS Number: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=d608216fe1e698d6b206b03588e6d6fe5ee661e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
Loss on drying ≤2 % (150°C)
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR