1-Heptanesulfonic acid sodium salt 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 1-heptanesulfonateSodium 1-heptanesulphonateSodium heptane-1-sulfonateSodium heptane-1-sulphonate

20613-0250 AC20613-0050 20613-1000 20613-5000
TS20613-5000EA 1543 USD
TS20613-5000 TS20613-1000 TS20613-0050 TS20613-0250
1-Heptanesulfonic acid sodium salt 98%
1-Heptanesulphonic acid sodium salt
Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://us.vwr.com/stibo/web/std.lang.all/07/91/2300791.jpg

Order Now

Specification Test Results

Appearance White Powder or flakes
Infrared spectrum Conforms
Loss on drying ≤9 % (100°C, 3 h) (vacuum)
Solubility (10 % in water) Clear colorless
Titration after Ion exchange ≥97.5 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR