Ethylenediamine 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Diaminoethane1,2-Ethanediamine

22042-0010 22042-2500 22042-0025
TS22042-0025EA 143 USD
TS22042-0025 TS22042-2500 TS22042-0010
Ethylenediamine 99+%
Ethylenediamine
Formula: C₂H₈N₂
MW: 60.1 g/mol
Boiling Pt: 117…118 °C (760 mmHg)
Melting Pt: 11 °C
Density: 0.898 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8,II
Merck Index: 13,03829
https://us.vwr.com/stibo/web/std.lang.all/99/69/2299969.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4555 to 1.4575 (20°C, 589 nm)
Water ≤0.5 % (Coulometric)
GC ≥99.0 %
Color scale ≤30 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR