EDTA disodium salt dihydrate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethylenediaminetetraacetic acid disodium dihydrate
, Ethylenediaminetetra-acetic acid disodium salt dihydrate
, Edetate disodium dihydrate
, Ethylenediaminetetraacetic acid, disodium salt, dihydrate
, Ethylenediaminetetraacetic acid disodium salt dihydrate
, Kestranal® 2S 2 H2O
, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate
, Titriplex III
, Kestranal® 2S dihydrate
, Edetic acid disodium salt dihydrate
, Disodium dihydrogen ethylenediaminetetraacetate dihydrate
, Sequestrene Na₂
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O MW: 372.24 g/mol Melting Pt: 252 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 EINECS: 205-358-3 Merck Index: 13,03543 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration Complexometric | 99.0 to 101.0 % |
pH | 4.0 to 6.0 (5 % at 25°C) |
Iron (Fe) | ≤100 ppm |
Heavy metals (as Pb) | ≤50 ppm |
Nitrilotriacetic acid | ≤0.1 % |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...