L(+)-Sodium lactate 99%, Extra Pure

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(+)-2-Hydroxypropionic acid sodium salt(S)-(+)-Lactic acid sodium salt(S)-(+)-2-Hydroxypropanoic acid sodium saltL(+)-Lactic acid sodium saltSodium (S)-2-hydroxypropanoateSodium-(S)-lactate

43922-0100 43922-0500
TS43922-0100EA 114 USD
TS43922-0100 TS43922-0500
L(+)-Sodium lactate 99%, Extra Pure
L(+)-Sodium lactate
Formula: C₃H₅NaO₃
MW: 112.06 g/mol
Melting Pt: 161.6…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00066576
CAS Number: 867-56-1

Order Now

Specification Test Results

Appearance White to light yellow Powder or solid
Infrared spectrum Conforms
Melting point 160 to 166 °C
Water ≤1.0 %
Specific optical rotation -12° to -13° (20 °C, 589 nm) (C=1 in H2O)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR