Silver sulfate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Disilver sulphateSilver(I) sulfateDisilver sulfate

AC19407-0100 AC19407-5000 19407-1000
TS19407-0100EA 127 USD
TS19407-0100 TS19407-5000 TS19407-1000
Silver sulfate ACS
Silver sulphate
Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085 °C (760 mmHg)
Melting Pt: 652 °C
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/94/57/2299457.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥98.0 %
Nitrate (NO3) ≤0.001 %
Iron (Fe) ≤0.001 %
Insoluble matter ≤0.02 % (in water and AgCl)
Subst not pptd by HCl ≤0.03 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR