Methanesulfonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

43297-0010 43297-0050
TS43297-0010EA 167 USD
TS43297-0010 TS43297-0050
Methanesulfonic acid 98%
Methanesulphonic acid
Formula: CH₃SO₃H
MW: 96.11 g/mol
Boiling Pt: 217
Melting Pt: 20
Density: 1.481 g/cm³
Flash Pt: 189 °C
Storage Temperature: Ambient
MDL Number: MFCD00007518
CAS Number: 75-75-2
UN: 3265
ADR: 8 III
Merck Index: 13,05981
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4285 to 1.4315 (20°C, 589 nm)
Titration with NaOH ≥97.5 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.2 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR