Methanesulfonic acid 99%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

12561-0010 12561-2500 12561-0050 12561-0051
TS12561-0010EA 231 USD
TS12561-0010 TS12561-2500 TS12561-0050 TS12561-0051
Methanesulfonic acid 99%, extra pure
Methanesulphonic acid
Formula: CH₃SO₃H
Boiling Pt: 167 °C (10 mmHg)
Melting Pt: 285…286 °C
Density: 1.481 g/cm³
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 205-290-4
UN: 3265
ADR: 8,III
Merck Index: 13,05981
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Titration with HClO4 99.0 to 100.5 % (On dried substance)
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR