Potassium-tert-butoxide 25% (w/w) 2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium tert-butanolatePotassium t-butoxidePotassium tert-butoxidePotassium tert-butylate

44583-1000 44583-8000
TS44583-8000EA 695 USD
TS44583-8000 TS44583-1000
Potassium-tert-butoxide 25% (w/w) 2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
Density: 0.86 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00012162
CAS Number: 865-47-4
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid (May develop some turbidity or precipitate)
Strength 1.90 to 2.20 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR