Potassium-tert-butoxide 98+%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium tert-butanolatePotassium t-butoxidePotassium tert-butoxidePotassium tert-butylate

16888-5000 AC16888-0250 AC16888-0025 AC16888-0050 AC16888-1000
TS16888-0025EA 456 USD
TS16888-0025 TS16888-1000 TS16888-0050 TS16888-5000 TS16888-0250
Potassium-tert-butoxide 98+%, pure
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
Melting Pt: 240…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II

Order Now

Specification Test Results

Appearance White to off-white Powder
Titration with HCl ≥98.0 %
KOH + K2CO3 (as KOH) ≤0.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR