Hydroxylammonium chloride ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

41205-0025 41205-1000 41205-5000
TS41205-0025EA 936 USD
TS41205-0025 TS41205-1000 TS41205-5000
Hydroxylammonium chloride ACS
Hydroxylammonium chloride
Formula: NH₂OH·HCl
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 13,04853

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Residue after ignition ≤0.05 %
Titration with KMnO4 ≥96.0 %
Clarity of alcohol solution Passes test
Ammonium (NH4) Passes test
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Sulfur compounds (as SO4) ≤0.005 %
Titrable free acid ≤0.25 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR