Tetraethyl orthosilicate (TEOS) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetraethoxysilane
15781-0010
15781-0025
15781-0100
AC15781-1000
AC15781-2500
TS15781-0010EA
107
USD
TS15781-0010
TS15781-0025
TS15781-0100
TS15781-1000
TS15781-2500
Tetraethyl orthosilicate (TEOS) 98%
Tetraethyl orthosilicate (TEOS)
Formula:
Si(OC₂H₅)₄ Boiling Pt: 166…169 °C (760 mmHg) Melting Pt: –77 °C Density: 0.94 g/cm³ Flash Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009062 CAS Number: 78-10-4 EINECS: 201-083-8 UN: 1292 ADR: 3,III Merck Index: 13,03882 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3820 to 1.3840 (20°C, 589 nm) |
GC | ≤1 % Ethanol |
Color scale | ≤20 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...