Dimethyl carbonate, extra dry 99+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

42873-0010 428731000
TS42873-0010EA 193 USD
TS42873-0010 TS428731000
Dimethyl carbonate, extra dry 99+%, AcroSeal™
Dimethyl carbonate
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 90
Melting Pt: 2…4 °C
Density: 1.069 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008420
CAS Number: 616-38-6
UN: 1161
ADR: 3 II
Merck Index: 12,06117

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Color scale ≤30 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR