N,N-Dimethylacetamide 99.5%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

37871-0025 378710200 378710010
TS37871-0025EA 154 USD
TS37871-0025 TS378710200 TS378710010
N,N-Dimethylacetamide 99.5%, Extra Pure
N,N-Dimethylacetamide
Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 164…166 °C (760 mmHg)
Melting Pt: –20 °C
Density: 0.937 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4370 to 1.4390 (20°C, 589 nm)
Water ≤0.1 %
GC ≥99.4 %
Color scale ≤10 APHA
Free acid (CH3COOH) ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR