Iron(II) chloride tetrahydrate 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ferrous chloride tetrahydrate
, Iron dichloride tetrahydrate
AC20508-0050
AC20508-0010
AC20508-0051
20508-2500
TS20508-2500EA
102.17
USD
TS20508-2500
TS20508-0010
TS20508-0050
TS20508-0051
Iron(II) chloride tetrahydrate 99+%
Iron (II) chloride tetrahydrate
Formula:
FeCl₂·4H₂O MW: 198.81 g/mol Melting Pt: 105…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149709 CAS Number: 13478-10-9 EINECS: 231-843-4 UN: 3260 ADR: 8,II |
Specification Test Results
Appearance | Yellow to green Crystalline powder, crystals and/or chunks |
Titration with KMnO4 | ≥99.0 % |
Sulfate (SO4) | ≤200 ppm |
Arsenic (As) | ≤5 ppm |
Copper (Cu) | ≤50 ppm |
Lead (Pb) | ≤50 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...