Hydrobromic acid 48% in water, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrogen bromide

12317-0010 12317-0025
TS12317-0025EA 251.84 USD
TS12317-0025 TS12317-0010
Hydrobromic acid 48% in water, pure
Hydrogen bromide
Formula: HBr
MW: 80.91 g/mol
Melting Pt: –11 °C
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow to orange Solution (May darken on storage)
Titration with NaOH ≥47 %
Melting point 62 to 66 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR