Imidazole 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Glyoxaline
, 1H-Imidazole
, Glyoxalamine
12202-0020
12202-1000
12202-5000
12202-0100
12202-0050
TS12202-0050EA
23.2
USD
TS12202-0050
TS12202-0020
TS12202-5000
TS12202-1000
TS12202-0100
Imidazole 99%
Imidazole
Formula:
C₃H₄N₂ MW: 68.08 g/mol Boiling Pt: 255…256 °C (760 mmHg) Melting Pt: 86…90 °C Flash Pt: 145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005183 CAS Number: 288-32-4 EINECS: 206-019-2 UN: 3263 ADR: 8,III Merck Index: 13,04935 |
Specification Test Results
Appearance | White to light yellow Flakes |
Infrared spectrum | Conforms |
Water | ≤0.2 % (K.F.) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...