Morpholine 99+%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
15868-0010
15868-0050
15868-0025
15868-0100
TS15868-0025EA
122
USD
TS15868-0025
TS15868-0010
TS15868-0100
TS15868-0050
Morpholine 99+%, Extra Pure
Morpholine
Formula:
C₄H₉NO MW: 87.12 g/mol Boiling Pt: 126…130 °C (760 mmHg) Melting Pt: –5 °C Density: 0.99 g/cm³ Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005972 CAS Number: 110-91-8 EINECS: 203-815-1 UN: 2054 ADR: 8,I |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4530 to 1.4550 (20°C, 589 nm) |
GC | ≥99.0 % |
Color scale | ≤20 APHA |
Insoluble matter | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...