Potassium hydrogen sulfate 99%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Potassium bisulfate, Potassium bisulphate
44761-5000
AC44761-0025
AC44761-0100
TS44761-5000EA
54
USD
TS44761-5000
TS44761-0025
TS44761-0100
Potassium hydrogen sulfate 99%, extra pure
Potassium hydrogen sulphate
Formula:
KHSO₄ Melting Pt: 214 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011404 CAS Number: 7646-93-7 UN: 2509 ADR: 8,II |
Specification Test Results
Appearance | White Crystalline powder |
Solubility | (10 % in water) Clear colorless |
Titration with NaOH | ≥98.5 % |
Chloride (Cl) | ≤0.002 % |
Ammonium (NH4) | ≤0.003 % |
Iron (Fe) | ≤0.003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...